Digital polarimeter, ADP430
Lieferant: BELLINGHAM STANLEY
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
37-30
BELL37-30EA 10651 EUR InStock
BELL37-30
Digital polarimeter, ADP430
Polarimeter
Single wavelength polarimeter with low maintenance LED light source. Suitable for sugar, food, chemical and pharmaceutical industries.
  • For use with waterbaths, ATC, or where no temperature control is required
  • Up to 200 mm tube length
  • Standard ICUMSA tubes
  • Continuous or single-shot readings
For a fully featured polarimeter without the need for internal temperature control, the ADP430 polarimeter comes complete with all the features of the ADP450 minus the XPC Technology. The ADP430 is ideal for users who prefer or already have waterbaths for temperature control or where temperature compensation or stability is not required.
These polarimeters contain proven optics featuring 'no maintenance' yellow LED and interference filter. Combined with a photodiode detector these technologies allow for the reading of samples of up to 3,0 Ext.Ø at the commonly used sodium (589 nm) wavelength. ADP400 polarimeters feature angular and sugar scales (ISS) and can be programmed with user scales as well as industry standard methods for displaying invert sugar, inversion (A to B) or when applying other factors such as tube length and concentration, Specific Rotation (or concentration when entering specific rotation).
ADP400 Series polarimeters have a resolution of 0,001 angular degrees (°A) and are capable of reading to an accuracy of ±0,010 °A. When working under laboratory controlled conditions, ADP400 polarimeters also offer excellent reproducibility of up to 0,002 °A.
You can take readings with the ADP400 Series polarimeter in either continuous measurement or our new ‘single-shot’ mode. In continuous mode a reading is taken over a set period of time and a stabilised result displayed at the end of the period. It is excellent for monitoring instrument and sample stability, giving the user a 'feel' for the reading obtained.
ADP400 Series polarimeters come equipped with a new full colour 4" high definition display meaning results are clearly visible to the operator. The expanded memory ensures that over 8000 measurements and recorded logs of instrument configuration can be saved and viewed or output to LIMS or Secure PDF.
Calibration and configuration of the instruments can be password protected, accessible by keypad entry or, for convenience, using a fully configurable RFID tag. This, together with the audit trail, facilitates operation in environments conforming to FDA regulation 21 CFR Part 11 or GLP. The ADP400 Series polarimeters incorporate a number of industry standard interfaces making it easy to connect to peripheral devices including barcode readers, printers and USB memory sticks for external storage.
Zertifizierungen: USP, EP, BP, JP, FDA 21 CFR Part 11 or GLP.
Bestellinformation: Quartz control plates can be ordered separately. Supplied with 200 mm glass centre fill polarimeter tube, pack of three RFID tags, instruction manual and certificate of conformity.
Lieferumfang: EU, UK and US plugs are supplied with the unit, CH and Danish plugs must be ordered separately.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge