Ring wrenches, BOLA
Lieferant: Bohlender
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
D701-24
BOHLD701-24EA 9.3 EUR InStock
BOHLD701-24
Ring wrenches, BOLA
Schraubenschlüssel
These PP ring wrenches can be used for opening and closing multiple distributors for barrels.
  • Very good chemical resistance
  • Temperature resistant from –20 to +110 °C
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge