Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-NOS3 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-0356R-CY7
BOSSBS-0356R-CY7EA 900 EUR InStock
BOSSBS-0356R-CY7
Anti-NOS3 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Digoxin is a beta blocker drug originally derived from the foxglove plant, Digitalis lanata. Digoxin is used primarily to improve the pumping ability of the heart in congestive heart failure (CHF), and treat problems such as high blood pressure. It is also used to help normalize some dysrhythmias (abnormal types of heartbeat).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NOS3
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Others

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge