Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-IFNB1 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-0787R-CY5.5
BOSSBS-0787R-CY5.5EA 909 EUR InStock
BOSSBS-0787R-CY5.5
Anti-IFNB1 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Interferon-Beta, IFN-Beta mainly by the fibroblast cell factor, belonging to a single gene encodes a protein that can interfere with the virus RNA or DNA replication and inhibit virus growth, and significantly enhance NK cell activity, through the promotion of MHC â…  molecule expression and enhance the CTL of the virus-infected cells role in the identification and destruction.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IFNB1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge