Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Keratocan Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11054R-A488
BOSSBS-11054R-A488EA 876 EUR InStock
BOSSBS-11054R-A488
Anti-Keratocan Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Small leucine-rich proteoglycans (SLRPs) such as Decorin, Biglycan, Fibromod-ulin, Keratocan, Lumican and Osteoglycin mediate extracellular matrix organization and are binding partners of TGF Beta. The Decorin core protein binds to growth factors, intercellular matrix molecules such as Fibronectin and Throm-bospondin, and to the Decorin endocytosis receptor. Fibromodulin is a collagen-binding keratan sulphate proteoglycan that influences adhesion processes of connective tissue and plays a role in fibrillogenesis by regulating collagen fibril spacing and thickness. Keratocan (KTN) develops corneal transparency and maintains the stromal matrix structure. Keratocan is a secreted protein in the extracellular matrix that binds to keratan sulfate chains. Keratocan is mainly detected in the cornea, but can also be expressed in trachea, intestine, ovary, lung and skeletal muscle. Defects in the gene encoding for Keratocan can cause cornea plana 2 (CNA2), an autosomal recessive disorder where the forward convex curvature of the cornea is flattened.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Keratocan
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge