Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-DOK6 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11066R-HRP
BOSSBS-11066R-HRPEA 900 EUR InStock
BOSSBS-11066R-HRP
Anti-DOK6 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
The downstream of kinase family (Dok-1-7) are members of a class of “docking” proteins that include the tyrosine kinase substrates IRS-1 and Cas, which contain multiple tyrosine residues and putative SH2 binding sites. Dok-4, Dok-5 and Dok-6 are more similar to each other than to the other Dok family members, and may constitute a subfamily of the DOK genes. Dok-5 is a tyrosine kinase substrate that enhances c-Ret-dependent activation of mitogen-activated protein kinase (MAPK). Dok-5 transcript is abundant in muscle and increases during T cell activation. Dok-5 protein undergoes tyrosine phosphorylation in response to insulin and insulin-like growth factor-1. Dok-6 is highly expressed in the developing central nervous system. It associates with Ret to transduce Ret-mediated processes such as axonal projection.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: DOK6
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge