Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-TMPRSS5 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11183R-A555
BOSSBS-11183R-A555EA 893 EUR InStock
BOSSBS-11183R-A555
Anti-TMPRSS5 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Extracellular proteases mediate the digestion of neighboring extracellular matrix components in initial tumor growth, allow desquamation of tumor cells into the surrounding environment, provide the basis for invasion of basement membranes in targeted metastatic organs and are required for release and activation of many growth and angiogenic factors. TMPRSS5 (transmembrane protease, serine 5), also known as spinesin, is a 457 amino acid single-pass type II membrane protein that is expressed specifically in brain and is thought to play a role in hearing. A member of the peptidase S1 family, TMPRSS5 contains one peptidase S1 domain and an SRCR domain, and is encoded by a gene that maps to human chromosome 11q23.2. Defects in the gene encoding TMPRSS5 are associated with deafness.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TMPRSS5
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge