Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-HOXA3 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11292R-A555
BOSSBS-11292R-A555EA 893 EUR InStock
BOSSBS-11292R-A555
Anti-HOXA3 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
The Hox proteins play a role in development and cellular differentiation by regulating downstream target genes. Specifically, the Hox proteins direct DNA-protein and protein-protein interactions that assist in determining the morphologic features associated with the anterior-posterior body axis. The mammalian HOX gene complex consists of 39 genes that are located on four linkage groups, which are dispersed over four chromosomes. HOX genes that occupy the same relative position along the 5’ to 3’ coordinate (trans-paralogous genes) are more similar in sequence and expression pattern than adjacent HOX genes on the same chromosome. HoxA3, in conjunction with Pax1, mediates the development of the thymus, parathyroid gland, and carotid body. Its expression in the third pharyngeal arch and pouch is required for development of the third arch artery, and homozygous null HoxA3 mutants lack the carotid body. HoxA3 also regulates hindbrain development by controlling the axon projection pattern of motor neurons and sensory neurons of the proximal and distal ganglia.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HOXA3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge