Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Neuroglycan C Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11323R-CY5
BOSSBS-11323R-CY5EA 900 EUR InStock
BOSSBS-11323R-CY5
Anti-Neuroglycan C Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Neuroglycan C is a brain-specific chondroitin sulfate proteoglycan (CSPG) implicated in the proliferation of neural stem and progenitor cells. Neuro-glycan C is a single-pass membrane protein that can manifest as a part-time proteoglycan depending on the tissue expressing it. In its proteoglycan form, Neuroglycan C exhibits chondroitin sulfate glycans and functions as a receptor for midkine, a growth factor that binds heparin, to affect cytoskeletal changes. By means of ectodomain shedding, the ectodomain of Neuroglycan C is able to enhance neurite outgrowth from neurons. Neurite growth stimulation is affected by both an EGF-like and an acidic amino acid domain found on the shed ectodomain. Both domains instigate neurite growth, however, these domains exhibit differing functionality as to number of neurites produced and neuron types stimulated.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: Neuroglycan C
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge