Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-APBA1 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11334R-CY7
BOSSBS-11334R-CY7EA 900 EUR InStock
BOSSBS-11334R-CY7
Anti-APBA1 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
The Beta-Amyloid precursor protein (Beta-APP) is a major constituent of the amyloid deposits in patients with Alzheimer’s disease. The Beta-Amyloid precursor is known to interact with several proteins, including X11 and the G heterotrimetric protein APP-BP1. The neuronal, transmembrane protein X11 is known to bind to the ∫-Amyloid precursor protein via a phosphotyrosine binding (PTB) domain, reducing the secretion of cellular Beta-APP and slowing Beta-APP processing pathways. X11 binds specifically to the YENPTY motif, which is involved in the internalization of Beta-APP. Multiple splice varitents of X11 have been identified, including X11å (also designated Mint 1), X11Beta (Mint 2) and X11(Mint 3).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: APBA1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge