Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-BEGAIN Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11335R-FITC
BOSSBS-11335R-FITCEA 900 EUR InStock
BOSSBS-11335R-FITC
Anti-BEGAIN Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
BEGAIN is a 593 amino acid protein that localizes to cytoplasm and membrane. BEGAIN interacts with PSD-95 and SAPAP1 and forms a ternary complex and may sustain the structure of the postsynaptic density (PSD). BEGAIN is a novel PSD component associated with the core complex of PSD-95 and SAPAP. Because BEGAIN and SAPAP interact with the same region of PSD-95, BEGAIN and SAPAP may compete for the binding to PSD-95 and cannot interact with PSD-95 simultaneously. The C-terminal region of BEGAIN is involved in the interaction with PSD-95 whereas the N-terminal region has a coiled-coil structure that may interact with other molecules. BEGAIN is specifically expressed in brain and enriched in the PSD fraction. BEGAIN is also expressed in neurons and enriched at synaptic junctions, and is likely involved in the organization of synaptic junction components.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: BEGAIN
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge