Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CASK Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11338R-CY5
BOSSBS-11338R-CY5EA 900 EUR InStock
BOSSBS-11338R-CY5
Anti-CASK Rabbit Polyclonal Antibody (Cy5®)
Antikörper
The MAGUK (membrane-associated guanylate kinase homologs) family of proteins contain multiple protein-binding domains and are involved in cell junction organization, tumor suppression, and signaling. CASK (also designated LIN-2) belongs to a MAGUK subfamily which is characterized by a novel domain structure that consists of a calcium/calmodulin- dependent protein kinase domain followed by PDZ, SH3 and guanylate kinase-like (GUK) domains. CASK is expressed in rat brain where it binds to cell-surface proteins, such as neurexin and syndecan, and is thought to be involved in signaling at neuronal synapses. CASK translocates to the nucleus and interacts with Tbr-1 to form a complex, which binds to a specific DNA sequence (the T-element), and induces the expression of specific genes, including Reelin. CASK displays a transcription regulation function, which appears crucial for cerebrocortical development.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CASK/CAMGUK
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge