Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SYNJ1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11376R-A488
BOSSBS-11376R-A488EA 893 EUR InStock
BOSSBS-11376R-A488
Anti-SYNJ1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
The inositol polyphosphate 5-phosphatases selectively remove the phosphate from the 5-position of various phosphatidylinositols, which generate second messengers in response to extracellular signals. Synaptojanins are characterized by an N-terminal SAC1-like sequence, a central 5-phosphate domain, and a unique C-terminal sequence and have been shown to use phosphatidylinositol 4,5-bisphosphate as a substrate. Synaptojanins exist as two isoforms, synaptojanin 1 and 2, which differ in the C-terminal domain, and each isoform has multiple variants produced by alternative splicing. Synaptojanin 1 is expressed as two major forms: the shorter is found in brain while the longer is expressed in peripheral tissues. Eight splice variants of synaptojanin 2 have been detected, including a brain specific isoform. Synaptojanins are thought to participate in the endocytosis of synaptic vesicles and the regulation of the actin cytoskeleton.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SYNJ1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge