Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CDKL5 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11538R-CY3
BOSSBS-11538R-CY3EA 900 EUR InStock
BOSSBS-11538R-CY3
Anti-CDKL5 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Cell cycle progression is controlled in part by a family of cyclin proteins and cyclin-dependent kinases (Cdks). Cdk proteins work in concert with the cyclins to phosphorylate key substrates involved in each phase of cell cycle progression. Another family of proteins, Cdk inhibitors, also plays a role in regulating the cell cycle by binding to cyclin-Cdk complexes and modulating their activity. CDKL5 (cyclin-dependent kinase-like 5) is a 1030 amino acid protein that belongs to the CMGC Ser/Thr protein kinase family. Expressed in brain, lung, kidney, prostate, ovary, placenta, pancreas and testis, CDKL5 is thought to play a role in cell cycle regulation. Defects in CDKL5 are a cause of several disorders, such as X-linked infantile spasm syndrome and Rett syndrome.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CDKL5
Clonality: Polyclonal
Clone: 0
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge