Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-EN2 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11552R-CY7
BOSSBS-11552R-CY7EA 883 EUR InStock
BOSSBS-11552R-CY7
Anti-EN2 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
The engrailed-2 gene, EN2, a murine homolog of the Drosophila homeobox gene engrailed (EN), is required for midbrain and cerebellum development and dorsal/ventral patterning of the limbs as well as apical ectodermal ridge formation. In Drosophila, the EN gene plays an important role during development in segmentation, where it is required for the formation of posterior compartments. Human EN-1 and EN-2 are homeodomain-containing proteins and have been implicated in the control of pattern formation during development of the central nervous system. Different mutations in the mouse homologs, EN-1 and EN-2, produce different developmental defects that frequently are lethal. EN-1 is highly expressed by essentially all dopaminergic neurons in the substantia nigra and ventral tegmentum. EN-1 and EN-2 regulate expression of a-synuclein, a gene that is genetically linked to Parkinson’s disease. During early brain development mouse EN-2 is expressed in a broad band across most of the mid-hindbrain region. EN-2 is also expressed in mouse myoblasts and has been assiciated with cerebellar hypoplasia.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: EN2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge