Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GSN Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-1160R-CY5.5
BOSSBS-1160R-CY5.5EA 900 EUR InStock
BOSSBS-1160R-CY5.5
Anti-GSN Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Gelsolin is a calcium dependent actin binding protein. It is a potent modulator of actin filament length and gelation. Gelsolin has been shown to exist in at least two variant forms, cytoplasmic gelsolin and plasma gelsolin. Plasma gelsolin has also been called actin depolymerizing factor (ADF) or brevin. Human plasma and rabbit macrophage gelsolins differ by the presence of a 25 amino acid residue extension at the NH2 terminus in the human plasma gelsolin, which appears to account for the difference in relative molecular weights.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Gelsolin
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge