Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-APPBP2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11639R-A488
BOSSBS-11639R-A488EA 893 EUR InStock
BOSSBS-11639R-A488
Anti-APPBP2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
APPBP2 is a hydrophilic, microtubule binding protein that functions in the trafficking of Beta-Amyloid precursor protein. It is expressed in a variety of cell types and localizes to the cytoplasm. APPBP2 shares homology with kinesin light chain. It consists of a coiled-coil domain, PKC phosphorylation sites, four imperfect C-terminal tandem repeats, eight tetratricopeptide repeats and N- and C-terminal globular structures. APPBP2 recognizes and binds to the basolateral sorting sequence (BaSS) present in the cytoplasmic domain of the Beta-Amyloid precursor protein. In addition, APPBP2 interacts with the androgen receptor and suppresses androgen signaling.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: APPBP2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge