Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SPG3A Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11759R-A488
BOSSBS-11759R-A488EA 893 EUR InStock
BOSSBS-11759R-A488
Anti-SPG3A Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Atlastins are Golgi-localized, integral membrane proteins that function as GTPases. The Atlastin proteins, also designated SPG3A and guanylate-binding protein 3, comprise a Dynamin superfamily that plays a role in axonal maintenance. Hereditary spastic paraplegia (HSP) is an inherited neurodegenerative disorder that is characterized by retrograde axonal degeneration. HSP primarily affects long corticospinal neurons and causes spastic lower extremity weakness. Spastin, a microtubule (MT)-severing AAA ATPase, is a binding partner of Atlastin that is involved in membrane dynamics. This Spastin/Atlastin binding may be involved in the biochemical pathway that leads to HSP development. Mutations in the Atlastin gene (SPG3A) account for approximately 10% of all autosomal dominant HSPs, while mutations in the Spastin gene (SPG4) account for almost 40%.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SPG3A/Atlastin
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge