Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-DORFIN/RNF19 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11778R-HRP
BOSSBS-11778R-HRPEA 900 EUR InStock
BOSSBS-11778R-HRP
Anti-DORFIN/RNF19 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Dorfin is a multi-pass membrane, RING-IBR type, E3 ubiquitin-protein ligase. It is widely expressed with highest levels found in heart and ubiquitous expression throughout the central nervous system. Dorfin functions by accepting ubiquitin in the form of a thioester from UBCH7 and UBC8 and then transferring it to the targeted substrates. Dorfin is responsible for ubiquitylating synphilin-1, CaSR and mutant variants of SOD-1, a protein at fault for familial ALS (amyotrophic lateral sclerosis). Dorfin physically interacts with VCP (Valosin-containing protein) via its C-terminus. Together these two proteins are associated with the formation of ubiquitylated inclusions (UBIs) that characterize many neurodegenerative disorders, such as Parkinson’s disease and ALS. This association with UBIs suggests that Dorfin plays an important role in the disease process.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: DORFIN/RNF19
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge