Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SLCO3A1 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11802R-CY3
BOSSBS-11802R-CY3EA 893 EUR InStock
BOSSBS-11802R-CY3
Anti-SLCO3A1 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
The organic anion transporter family of proteins mediate hepatic uptake of cardiac glycosides. OATP-D is a 710 amino acid member of the organic anion transporter protein family. As a multi-pass membrane protein, OATP-D mediates the Na+-independent transport of vasopressin, prostaglandins (PG) E1 and E2, thyroxine (T4), deltorphin II and other organic anions, but not estrone-3-sulfate, DPDPE, taurocholate, DHEAS or digoxin. OATP-D is ubiquitously expressed with highest levels present in leukocytes and spleen. OATP-D is expressed as four isoforms produced by alternative splicing events.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: SLCO3A1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge