Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-LHX5 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11870R-A488
BOSSBS-11870R-A488EA 893 EUR InStock
BOSSBS-11870R-A488
Anti-LHX5 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
During development, genetically distinct subtypes of motor neurons express unique combinations of LIM-type homeodomain factors, which regulate cell migration and guide motor axons to establish the fidelity of a binary choice in axonal trajectory. The LIM gene family encodes a set of gene products, which carry the LIM domain, a unique cysteine-rich zinc-binding domain. At least 40 members of this family have been identified in vertebrates and invertebrates, and are distributed into 4 groups according to the number of LIM domains and to the presence of homeodomains and kinase domains. The human LHX5 gene maps to chromosome 12q23-q24 and encodes a 402 amino acid protein. The hippocampus contains the neural circuitry, which is crucial for cognitive functions such as learning and memory. LHX5 regulates precursor cell proliferation and neuronal differentiation and migration during hippocampal development.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LHX5
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge