Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-LIMK1 + LIMK2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11871R-A350
BOSSBS-11871R-A350EA 893 EUR InStock
BOSSBS-11871R-A350
Anti-LIMK1 + LIMK2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
LIMK 1 and 2 likely regulate aspects of the cytoskeleton, through control of the organization of actin filaments. They can phosphorylate an actin-binding protein, cofilin which binds to actin monomers and polymers and promotes the disassembly of actin filament.The phosphorylation of cofilin via LIMK inactivates this potential. LIMK1 is highly active in the brain and spinal chord, where it is believed to be involved in the development of nerve cells whilst LIMK2 is ubiquitously expressed in many adult tissues. LIMK1 may play an important role in areas of the brain that are responsible for processing visual-spatial information (visuospatial constructive cognition). These parts of the brain are important for visualizing an object as a set of parts and performing tasks such as writing, drawing, constructing models, and assembling puzzles. LIMK1 is specifically stimulated by Rac, one of the Rho family proteins, while LIMK2 activity is activated under the control of other Rho family members, Rho and Cdc42, suggesting that two distinct pathways exist in the Rho family driven actin cytoskeleton dynamics.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LIMK1 + LIMK2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge