Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PTPRR Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11924R-A488
BOSSBS-11924R-A488EA 893 EUR InStock
BOSSBS-11924R-A488
Anti-PTPRR Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
The protein tyrosine phosphatase (PTP) family of proteins are signaling molecules that regulate processes such as cell growth, cell differentiation, oncogenic transformation and the mitotic cycle. PCPTP1, also known as PTPRR (Receptor-type tyrosine-protein phosphatase R), ECPTP, PTPBR7 or PTPRQ, is a 657 amino acid protein that functions to sequester inactive mitogen-activated protein kinases (MAPKs) to the cytoplasm. Expressed primarily in the brain with weaker expression in other parts of the body, PCPTP1 is a receptor-like molecule that is able to dephosphorylate MAPKs, thereby rendering them inactive. Three isoforms of PCPTP1 exist and are designated alpha, beta and gamma. The alpha form is localized to the cell membrane, while the beta and gamma forms are localized to the perinuclear areas within the cytoplasm.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: PTPRR
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge