Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SLC5A3 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11954R-A647
BOSSBS-11954R-A647EA 893 EUR InStock
BOSSBS-11954R-A647
Anti-SLC5A3 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Myo-inositol is involved in many important aspects of cellular regulation including membrane structure, signal transduction and osmoregulation. It is taken up into cells by the sodium/myo-inositol cotransporter (SMIT). SMIT activity maintains intracellular concentrations of myo-inositol; it is upregulated in response to hypertonic stress. The human SMIT protein is encoded by the SLC5A3 gene, which maps to chromosome 21q22.12. It is expressed in many human tissues, such as brain, kidney and placenta. Specifically, SMIT is abundantly expressed throughout the whole brain and spinal cord in fetal rat, but is downregulated in adult rat brain with the exception of the choroid plexus, where SMIT expression remains high. In kidney, SMIT localizes to the baso-lateral membranes of the thick ascending limb of Henle (TAL) and the inner medullary collecting duct (IMCD). Impaired SMIT activity is implicated in the pathogenesis of diabetes and Down syndrome.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: SLC5A3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge