Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Serine/threonine-protein kinase MAK-V Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11990R-A680
BOSSBS-11990R-A680EA 890 EUR InStock
BOSSBS-11990R-A680
Anti-Serine/threonine-protein kinase MAK-V Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
The HUNK (hormonally upregulated Neu-associated kinase) protein, also designated MAK-V in mouse, has been identified as a novel SNF1-related serine/threonine kinase. The human HUNK gene localizes to chromosome 21q22 and encodes a protein with nucleocytoplasmic distribution and localizes to the centrosome. Overexpression of the HUNK protein associates with approximately 50% of breast carcinomas, and may provide diagnostic-prognostic value as a molecular marker. Serine/threonine-protein kinase SNF1-like kinase 2 (SIK) phosphorylates Ser-794 of IRS1 in insulin-stimulated adipocytes, which may modulate the efficiency of insulin signal transduction. SIK is activated by phosphorylation on Thr-175 by STK11 in complex with STE20-related adapter- and CAB39.

Type: Primary
Antigen: Serine/threonine-protein kinase MAK-V
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 71-115/714
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge