Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Phosphoserine/threonine Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11994R
BOSSBS-11994REA 714 EUR InStock
BOSSBS-11994R
Anti-Phosphoserine/threonine Rabbit Polyclonal Antibody
Antikörper
A hallmark of signal transduction pathways is the reversible phosphorylation of serine and threonine residues within specific sequences, or motifs, in target proteins. Specific signaling motifs include not only sequences that are recognized by protein kinases, but also those that are recognized by phosphorylation-dependent binding proteins like 14-3-3. These modular phosphoprotein interacting domains are critical elements in modulating, directing and amplifying intracellular communications. Many critical protein kinases can be regulated by phosphorylation at a specific serine or threonine surrounded by phenylalanine or tyrosine. For example, Akt, an important kinase that regulates cell survival, is activated by phosphorylation at Ser473, a site surrounded by phenylalanine and tyrosine. RSK1, p70 S6 K, and certain PKC isoforms also contain a similar consensus phosphorylation site. Phosphorylation of these sites is required for kinase activity. The (Ser/Thr) Phe Antibody is a powerful tool for discovery of new proteins containing this important regulatory motif.

IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: Phosphoserine/threonine
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Others

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge