Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Paralemmin 1 Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12035R
BOSSBS-12035REA 725 EUR InStock
BOSSBS-12035R
Anti-Paralemmin 1 Rabbit Polyclonal Antibody
Antikörper
Paralemmin, also called Paralemmin-1 or PALM, is a widely expressed peripheral membrane protein that is involved in cell structure and shape. A hydrophobic protein, Paralemmin is anchored to the cytoplasmic side of the cell membrane via di-palmitoylation and prenylation of its C-terminal cysteine cluster. Functioning at the synapse to regulate neuronal plasticity and plasma membrane dynamics, Paralemmin can bind to the dopamine receptor D3, thereby reducing D3 expression and subsequent adenylate cyclase activity. Overexpression of Paralemmin induces fibroblasts to extend long filopodia and to assume extreme cell shapes, suggesting involvement in the formation and stabilization of the plasma membrane. Two isoforms of Paralemmin exists due to alternative splicing events.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: Paralemmin 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge