Anti-CNG4/GARP Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Lieferant: Bioss
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
BS-12093R-HRP
BOSSBS-12093R-HRPEA 900 EUR InStock
BOSSBS-12093R-HRP
Anti-CNG4/GARP Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Cyclic nucleotide-gated (CNG) cation channels are heteromeric complexes made up of principal alpha and modulatory beta subunits (1,2). The alpha subunits consist of CNG1-3 and form functional cation channels by themselves (1,2). The beta subunits consist of CNG4-6 and, unlike the alpha subunits, do not form functional channels, but rather modify the properties of channels (1,2). CNG channels are essential components of olfactory and visual transduction (1,2). In olfactory neurons, CNG2, CNG4.3 and CNG5 form Ca2+ permeable channels, which open and depolarize the cell in response to cAMP (1-3). In rod photoreceptors, CNG1 and CNG4.1 combine to form Ca ion permeable channels, which give rise to a current in response to cGMP (1-3). CNG3 and CNG6 are expressed in cone receptors and may combine to form a native cGMP-activated channel (2,3). CNG channels have been implicated in other areas (4-6). CNG1 is also expressed in medium-sized and small-sized arteries, suggesting a role for CNG in the regulation of arterial blood pressure and of blood supply to different regions (4). CNG1, CNG4.1 and CNG4.2 have been detected in the rat pineal gland (5). CNG2, CNG4.3 and CNG5 are present in GT1 cell lines and may play a role in the secretion of gonadotropin-releasing hormone (6).
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500
Type: Primary
Antigen: CNG4/GARP
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge