Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-UTF1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12207R-A350
BOSSBS-12207R-A350EA 893 EUR InStock
BOSSBS-12207R-A350
Anti-UTF1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
UTF1 is a 341 amino acid protein that localizes to the nucleus and is subject to post-translational phosphorylation. Associating with the TFIID complex via an interaction with the TATA box binding protein (TFIID), UTF1 binds to the N-terminal region of ATF-2 and, via this binding, acts as a transcriptional coactivator of ATF-2, thereby enhancing transcriptional activity. Human UTF1 shares 64% homology with its mouse counterpart, suggesting a similar role between species. The gene encoding UTF1 maps to human chromosome 10, which houses over 1,200 genes and comprises nearly 4.5% of the human genome. Defects in some of the genes that map to chromosome 10 are associated with Charcot-Marie Tooth disease, Jackson-Weiss syndrome, Usher syndrome, nonsyndromatic deafness, Wolman’s syndrome, Cowden syndrome, multiple endocrine neoplasia type 2 and porphyria.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: UTF1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge