Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-NAC1/BTBD14B Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12247R-A350
BOSSBS-12247R-A350EA 893 EUR InStock
BOSSBS-12247R-A350
Anti-NAC1/BTBD14B Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
NAC1 is an N-terminal homodimerization domain that contains multiple copies of kelch repeats and/or C2H2-type zinc fingers. Proteins that contain BTB domains are thought to be involved in transcriptional regulation via control of chromatin structure and function. BTBD14B (BTB/POZ domain-containing protein 14B), also known as NACC1 (nucleus accumbens associated 1), BEND8 or NAC1, is a 527 amino acid protein that localizes to both the nucleus and the cytoplasm and contains one BTB (POZ) domain. Existing as a homooligomer that interacts with HDAC3 and HDAC4, BTBD14B functions as a transcriptional repressor that influences the transcriptional activity of CRIF1 and is required for proteasome recruitment to the nucleus and cytoplasm in dendritic spines. BTBD14B is overexpressed in multiple carcinomas, suggesting a role in tumor development and metastasis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NAC1/BTBD14B
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge