Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PTCHD2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12324R-FITC
BOSSBS-12324R-FITCEA 900 EUR InStock
BOSSBS-12324R-FITC
Anti-PTCHD2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
PTCHD2 is a 1,392 amino acid multi-pass membrane protein that contains one SSD (sterol-sensing) domain and belongs to the patched family. Expressed in retina, brain and testis, PTCHD2 localizes to endoplasmic reticulum and colocalizes with cholesterol. PTCHD2 overexpression leads to increased cholesterol levels, suggesting that PTCHD2 may play a role in cholesterol homeostasis. PTCHD2 is further hypothesized to act as a link between thyroid hormone and cholesterol metabolism. Existing as two alternatively spliced isoforms, PTCHD2 is thought to assist in the release of lipid-anchored secreted proteins and is encoded by a gene that maps to human chromosome 1p36.22.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: PTCHD2
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge