Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CK1g2/Casein Kinase 1 gamma 2 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12425R-CY5
BOSSBS-12425R-CY5EA 900 EUR InStock
BOSSBS-12425R-CY5
Anti-CK1g2/Casein Kinase 1 gamma 2 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Casein kinases are operationally defined by their preferential utilization of acidic proteins such as caseins as substrates. They can phosphorylate a large number of proteins. Participates in Wnt signaling.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CK1g2
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge