Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-EEF2K Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13058R-CY3
BOSSBS-13058R-CY3EA 929 EUR InStock
BOSSBS-13058R-CY3
Anti-EEF2K Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Eukaryotic elongation factor 2 kinase (EEF2k) previously known as Ca2+/calmodulin dependent protein kinase III, is an abundant cytoplasmic protein highly specific for elongation factor 2 (eEf2). Phosphorylation of eEF2 by eEF2 kinase on specific threonine residues results in the inactivation of eEF-2 and in termination of mRNA translation. The activity of eEF2 kinase is not only dependent upon Ca2+ ions, calmodulin (CaM) and insulin, but is also regulated both negatively and positively via phosphorylation by different protein kinases (AMPK, S6K1, p90 RSK). There is also evidence that eEF-2 phosphorylation is involved in the regulation of cell cycle progression, cellular differentiation, oogensis and malignant tumors.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: EEF2K
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge