Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-FBXO45 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13150R-FITC
BOSSBS-13150R-FITCEA 893 EUR InStock
BOSSBS-13150R-FITC
Anti-FBXO45 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Belonging to the F-box family of proteins, FBXO45 is a 286 amino acid protein that contains one C-terminal F-box domain. F-box proteins are critical components of the SCF (Skp1-CUL-1-F-box protein)-type E3 ubiquitin ligase complex and are involved in substrate recognition and protein recruitment for ubiquitination. They are members of a larger family of proteins that are involved in the regulation of a wide variety of cellular mechanisms, including the cell cycle, the immune response, signaling cascades and developmental processes. They function by targeting proteins, such as cyclins, cyclin-dependent kinase inhibitors, I˚B-å and ∫-catenin, for degradation by the proteasome after ubiquitination. Via its F-box domain, FBXO45 can directly interact with Skp1 p19 and CUL-1. FBXO45 has been shown to be an estrogen-induced gene.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FBXO45
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge