Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-FDPS Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13153R
BOSSBS-13153REA 725 EUR InStock
BOSSBS-13153R
Anti-FDPS Rabbit Polyclonal Antibody
Antikörper
FDPS is a 419 amino acid enzyme belonging to the FPP/GGPP synthetase family. Localized to cytoplasm and peroxisome, FDPS expression is regulated by phorbol esters and polyunsaturated fatty acids. FDPS assists in cholesterol biosynthesis, post-translational protein modifications and synthesis of steroid hormones in the isoprenoid pathway.FDPS catalyzes the formation of farnesyl diphosphate (FPP), a precursor for several classes of essential metabolites including sterols, dolichols, carotenoids, and ubiquinones. FDPS is inactivated by interferon-induced RSAD2, which may result in the disruption of lipid rafts at the plasma membrane. Existing as a homodimer, FDPS may have anti-viral effects when inactivated by RSAD2. Reduced activity of FDPS in liver may partly be the cause of Zellweger syndrome and neonatal adrenoleukodystrophy, both of which are known to be peroxisomal deficiency diseases.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: FDPS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge