Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-FOXF2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13201R-A350
BOSSBS-13201R-A350EA 893 EUR InStock
BOSSBS-13201R-A350
Anti-FOXF2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
The FOX family of transcription factors share a common DIUA binding domain termed a winged-helix or forkhead domain. Many FOX proteins play important roles in development, metabolism, cancer and aging. Development of the vertebrate gut is controlled by paracrine crosstalk between the endodermal epithelium and the associated splanchnic mesoderm. FOXF2 (forkhead box F2), also known as FKHL or FREAC2, is expressed in lung and placenta, and has been shown to transcriptionally activate several lung-specific genes. FOXF2 interacts with transcription factors TFIIB and TBP, and may be involved in regulating transcription in embryogenesis and pattern formation in multicellular organisms. FOXF2-deficient mice develop cleft palate and an abnormal tongue, which suggests that FOXF2 may be critical for palatogenesis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FOXF2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge