Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SRSF10 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13229R-CY3
BOSSBS-13229R-CY3EA 900 EUR InStock
BOSSBS-13229R-CY3
Anti-SRSF10 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
FUSIP1 is a member of the Serine/Arginine (SR) family of splicing factors. Members of the SR family all contain one or more RNA recognition motifs (RRM) and an SR-rich domain. SR factors are not only essential for constitutive splicing but also regulate splicing in a concentration-dependent manner by influencing the selection of alternative splice sites. Expressed in a variety of tissues with low expression in kidney, liver and heart, FUSIP1 localizes to the cytoplasm and nuclear speckles. In its dephosphorylated form (occurring during M phase of the cell cycle), FUSIP1 functions as a potent general repressor of pre-mRNA splicing and can interact with U1 SnRNP 70. In its phosphorylated form, FUSIP1 interacts with Tra-2∫ and, together, they may cooperate in the regulation of splicing. Four isoforms exist for FUSIP1. In neurons, FUSIP1 isoforms may act to either positively or negatively regulate alternative splicing.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SRSF10
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge