Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GABPA Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13261R-CY3
BOSSBS-13261R-CY3EA 900 EUR InStock
BOSSBS-13261R-CY3
Anti-GABPA Rabbit Polyclonal Antibody (Cy3®)
Antikörper
The transcription factor GA-binding protein (GABP) is composed of two subunits, the Ets-related GABP-alpha and a GABP-alpha-associated subunit, GABP beta. GABP alpha binds to a specific DNA sequence and GABP beta exists as b1 and b2 splice variants that differ in their C-termini. In primary neuronal cultures, GABP beta is expressed in both the cytoplasm and the nucleus, whereas GABP alpha is expressed mainly in the nucleus. GABP is constitutively expressed as either a GABP alpha beta heterodimer or a GABP alpha b heterotetramer, both of which can modify GABP-dependent transcription in vitro and in vivo. The GABP alpha beta tetrameric complex performs many different functions, such as stimulating transcription of the adenovirus E4 gene, differentially activating BRCA1 expression in human breast cell lines, potentiating Tat-mediated activation of long terminal repeat promoter transcription and viral replication in certain cell types, acting as a coordinator of mitochrondrial and nuclear transcription for cytochrome oxidase in neurons and assisting in the regulation of rpL32 gene transcription.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GABPA/NRF2A
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge