Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GALNT11/GalNAc-T11 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13271R-A680
BOSSBS-13271R-A680EA 890 EUR InStock
BOSSBS-13271R-A680
Anti-GALNT11/GalNAc-T11 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
The UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase (GalNAc-T) family of enzymes are substrate-specific proteins that catalyze the transfer of GalNAc (N-acetylgalactosamine) to serine and threonine residues onto various proteins, thereby initiating mucin-type O-linked glycosylation in the Golgi apparatus. GalNAc-T11 (Polypeptide N-acetylgalactosaminyltransferase 11), also known as UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 11, is a 608 amino acid protein that catalyzes glycosylation of Muc1, Muc4.1 and EA2, though it does not display enzymatic preference for erythropoitein. The N-terminal domain is involved in substrate binding and manganese coordination, while the C-terminal domain is involved in UDP-Gal binding and catalytic reaction. GalNAc-T11 is highly expressed in kidney tubules, though it is not expressed in glomeruli. There are two isoforms of GalNAc-T11 that are produced as a result of alternative splicing events.

Type: Primary
Antigen: GALNT11/GalNAc-T11
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 401-500/608
Host: Rabbit
Isotype: IgG
Reactivity: Human; Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge