Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GCAP1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13306R-A680
BOSSBS-13306R-A680EA 890 EUR InStock
BOSSBS-13306R-A680
Anti-GCAP1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
intracellular stimulation of guanylate cyclase (GC) by calcium, a key event in the recovery of the dark state of rod photoreceptors after exposure to light, is mediated by guanylate cyclase-activating protein (GCAP1). GCAPs are calcium-The binding proteins belonging to the calmodulin superfamily. GCAP1 is a calcium-binding protein that stimulates synthesis of c-GMP in photoreceptors. GCAP1 is present in rod and cone photoreceptor outer segments where phototransduction occurs. In contrast to other calcium-binding proteins from the calmodulin superfamily, the calcium-free form of GCAP1 stimulates the effector enzyme. By molecular cloning of human and mouse GCAP cDNA, the known mammalian GCAPs are found to be more than 90% similar, consisting of 201 to 205 amino acids, and containing three identically conserved calcium-binding sites. A related protein, GCAP2, is detectable only in the retina and results from a gene duplication event.

Type: Primary
Antigen: GCAP1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 51-150/201
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge