Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Glycogenin 1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13447R-A750
BOSSBS-13447R-A750EA 890 EUR InStock
BOSSBS-13447R-A750
Anti-Glycogenin 1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Glycogen synthesis is initiated by the autoglucosylation of Glycogenin-1. Specifically, Glycogenin-1 glucosylates itself to begin the synthesis of glycogen in mammalian skeletal muscle. It acts as the primer to which further glucose monomers may be added. All of the Glycogenin-1 molecules contain at least one glucosyl residue before autoglucosylation begins. The first step of the glycogen synthesis occurs when a glucose molecule from UDP-glucose binds to the hydroxyl group of Tyr 194 on the Glycogenin-1 molecule. Using its glucosyltransferase activity, Glycogenin-1 adds more glucoses, each one coming from UDP-glucose. The glycosylation process reaches a plateau when five new glucose residues have been added, at which point glycogen synthase (GS) takes over and further elongates the chain. Glycogenin-1 remains covalently attached to the reducing end of the glycogen molecule.

Type: Primary
Antigen: Glycogenin 1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 251-350/350
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge