Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GPR17 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13521R-CY5
BOSSBS-13521R-CY5EA 900 EUR InStock
BOSSBS-13521R-CY5
Anti-GPR17 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
G protein-coupled receptor 17, GPR17, also known as uracil nucleotide/cysteinyl leukotriene receptor or P2Y-like receptor (P2YL), is a 367 amino acid member of the G-protein coupled receptor 1 family of proteins. While GPR17 is expressed in kidney, heart and umbilical vein endothelial cells, it is expressed in the highest levels in the brain. Upon brain injury, the extracellular concentrations of nucleotides and cysteinyl leukotrienes (CysLTs), two families of endogenous signaling molecules, increase significantly at the site of damage. In some neurons, GPR17, a membrane receptor for uracil nucleotide and CysLTs, is upregulated as well, infiltrating the lesioned area. GPR17 is thought to play a role in mediating neuronal death, remodeling brain circuitries by microglia and initiating remyelination in damaged neurons. Two named isoforms of GPR17 exist as a result of alternative splicing events.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GPR17
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge