Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SLAP Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13668R
BOSSBS-13668REA 725 EUR InStock
BOSSBS-13668R
Anti-SLAP Rabbit Polyclonal Antibody
Antikörper
The Src homology 3 (SH3) region is a small protein domain of approximately 60 amino acids present in a large group of proteins. In general, it exists in association with catalytic domains, as in the nonreceptor protein-tyrosine kinases and phospholipase C-? within structural proteins, such as spectrin or Myosin; and in small adapter proteins, such as Crk and GRB2. SH3 domains are often accompanied by SH2 domains of 100 amino acids, which bind to tyrosine-phosphorylated regions of target proteins, frequently linking activated growth factors to putative signal transduction proteins. The functions of SH3 domains are not as well defined. Deletion or mutation of SH3 domains generally activate the transforming potential of nonreceptor tyrosine kinases, suggesting that SH3 mediates negative regulation of an intrinsic transforming activity. 3BP1 has been identified as a protein with a high affinity proline-rich binding site for the SH3 domain of c-Abl p120. A similar putative adapter protein, designated Slap, for Src-like adapter protein, has been cloned. Slap contains a single SH2 and SH3 domain that exhibits homology with those from members of the Src kinase family. The N- and C-termini, however, are unique.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: SLAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge