Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Cadherin 20 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13716R-CY5
BOSSBS-13716R-CY5EA 900 EUR InStock
BOSSBS-13716R-CY5
Anti-Cadherin 20 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
The cadherins are a family of Ca2+-dependent adhesion molecules that function to mediate cell-cell binding critical to the maintenance of structure and morphogenesis. Cadherins each contain a large extracellular domain at the N-terminus, which is characterized by a series of five homologous repeats, the most distal of which is thought to be responsible for binding specificity. Cadherin-20, also known as protocadherin gamma B4, is a member of the protocadherin gamma gene cluster. Expressed primariy in fibroblasts, cadherin-20 plays a critical role in neuronal cell-cell connections in the brain. Through its ability to mediate cell-cell adhesion, caderin-20 is thought to help the body respond to injury by regulating post-traumatic wound healing.

Type: Primary
Antigen: Cadherin 20
Clonality: Polyclonal
Clone:
Conjugation: Cy5
Public Immunogen Range: 301-400/801
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge