Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PCDHB10 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13725R-CY3
BOSSBS-13725R-CY3EA 900 EUR InStock
BOSSBS-13725R-CY3
Anti-PCDHB10 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Protocadherins are a large family of cadherin-like cell adhesion proteins that are involved in the establishment and maintenance of neuronal connections in the brain. There are three protocadherin gene clusters, designated a, b and g, all of which contain multiple tandemly-arranged genes. PCDHB10 (protocadherin b10), also known as PCHB10 or PCDH-b10, is an 800 amino acid protein that is one of 16 proteins in the protocadherin b cluster. Unlike the a and g gene clusters, whose genes are spliced to downstream constant-region exons during transcription, members of the b cluster (such as PCDHB10) do not use constant-region exons to produce mRNAs. As a result, each protocadherin b gene encodes the transmembrane, extracellular and short cytoplasmic domains of the protein. Localized to the cell membrane, PCDHB10 is a single-pass type I membrane protein that contains six cadherin domains.

Type: Primary
Antigen: PCDHB10
Clonality: Polyclonal
Clone:
Conjugation: Cy3
Public Immunogen Range: 351-450/800
Host: Rabbit
Isotype: IgG
Reactivity: Human; Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge