Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PCDH21 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13731R-FITC
BOSSBS-13731R-FITCEA 900 EUR InStock
BOSSBS-13731R-FITC
Anti-PCDH21 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Protocadherins are a large family of cadherin-like cell adhesion proteins that are involved in the establishment and maintenance of neuronal connections in the brain. There are three protocadherin gene clusters, designated alpha, beta and gamma, all of which contain multiple tandemly arranged genes. PCDH21 (protocadherin 21), also known as PRCAD, is an 859 amino acid single-pass membrane protein that localizes to the outer segments of photoreceptor cells and contains six cadherin domains. Existing as multiple alternatively spliced isoforms, PCDH21 functions as a calcium-dependent cell adhesion protein that is thought to be required for the structural integrity of photoreceptor cells and may be involved in the formation and maintenance of neuronal networks.

Type: Primary
Antigen: PCDH21
Clonality: Polyclonal
Clone:
Conjugation: FITC
Public Immunogen Range: 601-700/859
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge