Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-IRAK2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-1427R-FITC
BOSSBS-1427R-FITCEA 900 EUR InStock
BOSSBS-1427R-FITC
Anti-IRAK2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
IRAK or Interleukin-1 Receptor-associated Kinase 1, is one of two putative serine/threonine kinases that become associated with the interleukin-1 receptor (IL1R) upon stimulation. This protein is partially responsible for IL1-induced upregulation of the ubiquitous transcription factor NF-kappa B.

Type: Primary
Antigen: IRAK2
Clonality: Polyclonal
Clone:
Conjugation: FITC
Public Immunogen Range: 21-150/625
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge