Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-C5orf20 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-15199R-HRP
BOSSBS-15199R-HRPEA 900 EUR InStock
BOSSBS-15199R-HRP
Anti-C5orf20 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
C5orf20 is a intronless gene is specifically expressed in dendritic cells (DCs), which are potent antigen-presenting cells involved in activating naive T cells to initiate antigen-specific immune response. The encoded protein is localized mainly in the perinucleus. One of the alleles (A/T) of this gene, that causes premature translation termination at aa 117, has been associated with an increased prevalence of major depression in humans

Type: Primary
Antigen: C5orf20
Clonality: Polyclonal
Clone:
Conjugation: HRP
Public Immunogen Range: 131-244/244
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge