Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-HERC2 Rabbit Polyclonal Antibody (Cy5)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-15460R-CY5
BOSSBS-15460R-CY5EA 900 EUR InStock
BOSSBS-15460R-CY5
Anti-HERC2 Rabbit Polyclonal Antibody (Cy5)
Antikörper
HERC2 is a belongs to the HERC gene family that encodes a group of unusually large proteins, which contain multiple structural domains. All members have at least 1 copy of an N-terminal region showing homology to the cell cycle regulator RCC1 and a C-terminal HECT (homologous to E6-AP C terminus) domain found in a number of E3 ubiquitin protein ligases. Genetic variations in this gene are associated with skin/hair/eye pigmentation variability. Multiple pseudogenes of this gene are located on chromosomes 15 and 16.

Type: Primary
Antigen: HERC2
Clonality: Polyclonal
Clone:
Conjugation: Cy5
Public Immunogen Range: 51-200/4834
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge