Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-IGHD Rabbit Polyclonal Antibody (Cy5)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-15573R-CY5
BOSSBS-15573R-CY5EA 900 EUR InStock
BOSSBS-15573R-CY5
Anti-IGHD Rabbit Polyclonal Antibody (Cy5)
Antikörper
Ig delta chain C region is an allelic product of the human IGHD gene. The two known IGHD alleles, IGHD*01 and IGHD*02, respectively produce isoforms 1, a secreted protein, and 2, a single-pass type I membrane protein. A member of the adaptive immune system, IgD are monomers expressed by activated B cells. Containing 3 Ig-like (immunoglobulin-like) domains, IgD chain C is located on chromosome 14 within the human heavy chain locus, lying on the 3? side of the IgM chain C region from the V-D-J cassette. Polyadenylation at certain sites along the heavy chain locus likely effects the mechanism that determines the alternative splicing event which results in the expression of either IgD chain C or IgM chain C. Some studies have suggested that antigenic coactivation of IgD+ B cells can have a negative influence on bone resorption during infectious events.

Type: Primary
Antigen: IGHD
Clonality: Polyclonal
Clone:
Conjugation: Cy5
Public Immunogen Range: 51-150/384
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge